Assay Details
| Gene Symbol: Osgin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027950.1 | NP_082226.1 | 1 - 2 | 135 | 66 | |
| GenBank mRNA | BC006032.1 | - | 1 - 2 | 130 | 66 | |
| BC022135.1 | - | 1 - 2 | 135 | 5099665 | 66 | |
| BC110674.1 | - | 1 - 2 | 122 | 66 | ||
| BC132440.1 | - | 1 - 2 | 104 | 40130717 | 66 | |
| BC138192.1 | - | 1 - 2 | 104 | 8861211 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC022135.1, BC132440.1, BC138192.1, NM_027950.1 transcripts.
Set Membership: |
|

Osgin1
Genomic Map