Assay Details
| Gene Symbol: Osgin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027950.1 | NP_082226.1 | 3 - 4 | 380 | 85 | |
| GenBank mRNA | AB221676.1 | - | 2 - 3 | 207 | 85 | |
| AK005892.1 | - | 1 - 2 | 143 | 85 | ||
| BC022135.1 | - | 3 - 4 | 380 | 5099665 | 85 | |
| BC110674.1 | - | 3 - 4 | 367 | 85 | ||
| BC132440.1 | - | 3 - 4 | 349 | 40130717 | 85 | |
| BC138192.1 | - | 3 - 4 | 349 | 8861211 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osgin1
Genomic Map