Assay Details
| Gene Symbol: Osgin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027950.1 | NP_082226.1 | 4 - 5 | 569 | 68 | |
| GenBank mRNA | AB221676.1 | - | 3 - 4 | 396 | 68 | |
| AK005892.1 | - | 2 - 3 | 332 | 68 | ||
| BC006032.1 | - | 3 - 4 | 910 | 68 | ||
| BC022135.1 | - | 4 - 5 | 569 | 5099665 | 68 | |
| BC110674.1 | - | 4 - 5 | 556 | 68 | ||
| BC132440.1 | - | 4 - 5 | 538 | 40130717 | 68 | |
| BC138192.1 | - | 4 - 5 | 538 | 8861211 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osgin1
Genomic Map