Assay Details
| Gene Symbol: Mettl7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027853.2 | NP_082129.2 | 1 - 2 | 553 | 151 | |
| GenBank mRNA | AK002239.1 | - | 1 - 2 | 554 | 151 | |
| BC024898.1 | - | 1 - 2 | 438 | 4215792 | 151 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
metabolic process
methylation
rRNA (adenine-N6,N6-)-dimethyltransferase activity
molecular_function
protein C-terminal carboxyl O-methyltransferase activity
tRNA (guanine-N2-)-methyltransferase activity
methyltransferase activity
C-methyltransferase activity
N-methyltransferase activity
O-methyltransferase activity
S-methyltransferase activity
RNA methyltransferase activity
mRNA methyltransferase activity
tRNA methyltransferase activity
protein methyltransferase activity
2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
rRNA methyltransferase activity
rRNA (uridine-2'-O-)-methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
rRNA (adenine-N6-)-methyltransferase activity
DNA-methyltransferase activity
tRNA (guanine-N1-)-methyltransferase activity
rRNA (cytosine-C5-)-methyltransferase activity
selenocysteine methyltransferase activity
arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
lysine N-methyltransferase activity
protein-lysine N-methyltransferase activity
tRNA (uracil) methyltransferase activity
tRNA (guanine) methyltransferase activity
tRNA (adenine) methyltransferase activity
tRNA (cytosine) methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
tRNA (adenine-N1-)-methyltransferase activity
rRNA (adenine) methyltransferase activity
rRNA (cytosine) methyltransferase activity
rRNA (guanine) methyltransferase activity
rRNA (uridine) methyltransferase activity
transferase activity
protein C-terminal leucine carboxyl O-methyltransferase activity
1-phenanthrol methyltransferase activity
protein-arginine N5-methyltransferase activity
methylarsonite methyltransferase activity
dimethylarsinite methyltransferase activity
4,5-dihydroxybenzo(a)pyrene methyltransferase activity
1-hydroxypyrene methyltransferase activity
1-hydroxy-6-methoxypyrene methyltransferase activity
demethylmenaquinone methyltransferase activity
cobalt-precorrin-6B C5-methyltransferase activity
cobalt-precorrin-7 C15-methyltransferase activity
cobalt-precorrin-5B C1-methyltransferase activity
cobalt-precorrin-3 C17-methyltransferase activity
dimethylamine methyltransferase activity
hydroxyneurosporene-O-methyltransferase activity
tRNA (adenine-57, 58-N(1)-) methyltransferase activity
methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
trimethylamine methyltransferase activity
methanol-specific methylcobalamin:coenzyme M methyltransferase activity
monomethylamine methyltransferase activity
P-methyltransferase activity
Se-methyltransferase activity
2-phytyl-1,4-naphthoquinone methyltransferase activity
tRNA (uracil-2'-O-)-methyltransferase activity
tRNA (cytosine-2'-O-)-methyltransferase activity
phosphomethylethanolamine N-methyltransferase activity
tRNA (cytosine-3-)-methyltransferase activity
rRNA (cytosine-2'-O-)-methyltransferase activity
rRNA (cytosine-N4-)-methyltransferase activity
trihydroxyferuloyl spermidine O-methyltransferase activity
biological_process
metabolic process
methylation
rRNA (adenine-N6,N6-)-dimethyltransferase activity
molecular_function
protein C-terminal carboxyl O-methyltransferase activity
tRNA (guanine-N2-)-methyltransferase activity
methyltransferase activity
C-methyltransferase activity
N-methyltransferase activity
O-methyltransferase activity
S-methyltransferase activity
RNA methyltransferase activity
mRNA methyltransferase activity
tRNA methyltransferase activity
protein methyltransferase activity
2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
rRNA methyltransferase activity
rRNA (uridine-2'-O-)-methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
rRNA (adenine-N6-)-methyltransferase activity
DNA-methyltransferase activity
tRNA (guanine-N1-)-methyltransferase activity
rRNA (cytosine-C5-)-methyltransferase activity
selenocysteine methyltransferase activity
arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
lysine N-methyltransferase activity
protein-lysine N-methyltransferase activity
tRNA (uracil) methyltransferase activity
tRNA (guanine) methyltransferase activity
tRNA (adenine) methyltransferase activity
tRNA (cytosine) methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
tRNA (adenine-N1-)-methyltransferase activity
rRNA (adenine) methyltransferase activity
rRNA (cytosine) methyltransferase activity
rRNA (guanine) methyltransferase activity
rRNA (uridine) methyltransferase activity
transferase activity
protein C-terminal leucine carboxyl O-methyltransferase activity
1-phenanthrol methyltransferase activity
protein-arginine N5-methyltransferase activity
methylarsonite methyltransferase activity
dimethylarsinite methyltransferase activity
4,5-dihydroxybenzo(a)pyrene methyltransferase activity
1-hydroxypyrene methyltransferase activity
1-hydroxy-6-methoxypyrene methyltransferase activity
demethylmenaquinone methyltransferase activity
cobalt-precorrin-6B C5-methyltransferase activity
cobalt-precorrin-7 C15-methyltransferase activity
cobalt-precorrin-5B C1-methyltransferase activity
cobalt-precorrin-3 C17-methyltransferase activity
dimethylamine methyltransferase activity
hydroxyneurosporene-O-methyltransferase activity
tRNA (adenine-57, 58-N(1)-) methyltransferase activity
methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
trimethylamine methyltransferase activity
methanol-specific methylcobalamin:coenzyme M methyltransferase activity
monomethylamine methyltransferase activity
P-methyltransferase activity
Se-methyltransferase activity
2-phytyl-1,4-naphthoquinone methyltransferase activity
tRNA (uracil-2'-O-)-methyltransferase activity
tRNA (cytosine-2'-O-)-methyltransferase activity
phosphomethylethanolamine N-methyltransferase activity
tRNA (cytosine-3-)-methyltransferase activity
rRNA (cytosine-2'-O-)-methyltransferase activity
rRNA (cytosine-N4-)-methyltransferase activity
trihydroxyferuloyl spermidine O-methyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P685999 | AK002239.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P685999 | BC024898.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P685999 | NM_027853.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P685999 | AK002239.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P685999 | BC024898.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P685999 | NM_027853.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 599593 | AK153713.1 | genbank_mrna_mouse |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100798 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P685999 | AK002239.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P685999 | BC024898.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P685999 | NM_027853.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P685999 | AK002239.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P685999 | BC024898.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P685999 | NM_027853.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544173 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100798 | - | - |
| Whole Mouse Genome G4122A | A_52_P685999 | AK002239.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P685999 | BC024898.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P685999 | NM_027853.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P685999 | AK002239.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P685999 | BC024898.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P685999 | NM_027853.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140649 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100798 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08415561 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08415571 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08415570 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08415574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08563374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08748882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08266844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08415562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08450954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08353648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08415570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544178 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10373601 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373603 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373602 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416980_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373600 | - | - |
| Mouse Genome 430 2.0 Array | 1416980_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416980_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71664_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71664_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000023931.mm.1 | NM_027853.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023931.mm.1 | NM_027853.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023931.mm.1 | TC1000003169.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023932.mm.1 | NM_027853.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023932.mm.1 | NM_027853.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023932.mm.1 | TC1000003169.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023930.mm.1 | NM_027853.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023930.mm.1 | NM_027853.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023930.mm.1 | TC1000003169.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416980_at | - | - |
| Mouse Expression Set 430 | 1416980_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416980_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416980_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416980_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416980_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 378379 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6400333 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6400333 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006400333 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2707227 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2707227 | - | - |
| mouse-6 v1.1 (Illumina) | scl071664.1_311-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6400333 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl071664.1_311-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6400333 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2707227 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2707227 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl071664.1_311-S | - | - |

Mettl7b
Genomic Map