Assay Details
| Gene Symbol: C87436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243741.1 | NP_001230670.1 | 9 - 10 | 2362 | 143 | |
| NM_001243742.1 | NP_001230671.1 | 9 - 10 | 1928 | 143 | ||
| NM_146170.4 | NP_666282.3 | 9 - 10 | 1869 | 143 | ||
| GenBank mRNA | AK047127.1 | - | 9 - 10 | 1870 | 143 | |
| AK077503.1 | - | 9 - 10 | 2249 | 143 | ||
| AK170524.1 | - | 10 - 11 | 1760 | 143 | ||
| AK171511.1 | - | 10 - 11 | 1760 | 143 | ||
| BC025632.1 | - | 9 - 10 | 1836 | 5321669 | 143 | |
| BC026926.1 | - | 10 - 11 | 1993 | 3586097 | 143 | |
| BC031913.1 | - | 9 - 10 | 1882 | 5376246 | 143 | |
| BU613064.1 | - | 5 - 6 | 587 | 143 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610260 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610261 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104583 | AK040219.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104583 | BC031913.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104583 | BF460653.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104583 | BC026926.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104583 | AK170524.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510229 | AF513359.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510229 | AK010660.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510229 | AK154680.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510229 | AK163587.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510229 | AK170151.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510229 | AK170440.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510229 | BC004777.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510229 | NM_133754.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 56618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67270 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 25390 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72508 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583730 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 71542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 280222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80458 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 799696 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425541 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 512604 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 775959 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 964808 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 956400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19617 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 684907 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138791 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677451 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786343 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729852 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125405 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 36278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 101678 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128970 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170942 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104583 | AK040219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104583 | BC031913.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104583 | BF460653.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104583 | BC026926.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104583 | AK170524.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05968941 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170942 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05968978 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05968962 | - | - |
| Whole Mouse Genome G4122A | A_51_P477627 | BC031913.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P477627 | BF460653.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P477627 | BC026926.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P477627 | BC025632.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P477627 | AK170524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P532841 | BC031913.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P532841 | BC026926.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P532841 | BC025632.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P532841 | AK170524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P532841 | AK171511.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P108397 | BC031913.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P108397 | BC026926.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P108397 | BC025632.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P108397 | AK170524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P108397 | AK171511.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137128 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05968941 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170942 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05049020 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05049021 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05049023 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610260 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05049022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05690360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05401097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05136394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05690362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05302002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05708721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05690358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05136400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05560927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05203113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05371244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05665846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05866832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05049023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05785605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05708723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05147675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05690371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05440772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05168382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05049020 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10539738 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539731 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539735 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539734 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539733 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539736 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539729 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539732 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539730 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539728 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425776_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458430_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539727 | - | - |
| Mouse Genome 430 2.0 Array | 1457972_at | - | - |
| Mouse Genome 430 2.0 Array | 1425776_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425776_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1458430_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457972_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458430_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232196_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232196_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600007714.mm.1 | NM_001243741.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007714.mm.1 | NM_001243741.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007714.mm.1 | NM_001243742.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007714.mm.1 | NM_001243742.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007714.mm.1 | NM_146170.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007714.mm.1 | NM_146170.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007714.mm.1 | TC0600000989.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600007713.mm.1 | NM_001243741.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007713.mm.1 | NM_001243741.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007713.mm.1 | NM_001243742.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007713.mm.1 | NM_001243742.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007713.mm.1 | NM_146170.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007713.mm.1 | NM_146170.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007713.mm.1 | TC0600000989.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425776_a_at | - | - |
| Mouse Expression Set 430 | 1425776_PM_a_at | - | - |
| Mouse Expression Set 430 | 1457972_at | - | - |
| Mouse Expression Set 430 | 1458430_at | - | - |
| Mouse Expression Set 430 | 1457972_PM_at | - | - |
| Mouse Expression Set 430 | 1458430_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425776_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425776_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425776_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425776_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457972_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457972_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458430_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458430_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2940288 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002940288 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229035 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229035 | - | - |
| mouse-6 v1.1 (Illumina) | scl29800.9.1_6-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2940288 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29800.9.1_6-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2940288 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2882473 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1229035 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2882473 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1229035 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl29800.9.1_6-S | - | - |

C87436
Genomic Map