Assay Details
| Gene Symbol: Syp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009305.2 | NP_033331.2 | 1 - 2 | 149 | 64 | |
| GenBank mRNA | AK034291.1 | - | 1 - 2 | 92 | 64 | |
| AK043756.1 | - | 1 - 2 | 46 | 64 | ||
| AK090259.1 | - | 1 - 2 | 46 | 64 | ||
| BC052354.1 | - | 1 - 2 | 43 | 64 | ||
| BY239748.1 | - | 1 - 2 | 64 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Syp
Genomic Map