Assay Details
| Gene Symbol: Homer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146153.1 | NP_001139625.1 | 8 - 9 | 969 | 101 | |
| GenBank mRNA | AF093261.1 | - | 6 - 7 | 796 | 101 | |
| BU054773.1 | - | 6 - 7 | 673 | 101 | ||
| BU707796.1 | - | 3 - 4 | 397 | 101 | ||
| CN539202.1 | - | 6 - 7 | 667 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Homer3
Genomic Map