Assay Details
| Gene Symbol: Arhgef9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK163874.1 | - | 1 - 2 | 100 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK163874.1 transcripts.
Set Membership: |
|

Arhgef9
Genomic Map