Assay Details
| Gene Symbol: Acp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007387.2 | NP_031413.1 | 9 - 10 | 978 | 73 | |
| GenBank mRNA | AK089401.1 | - | 9 - 10 | 969 | 73 | |
| AK128963.1 | - | 9 - 10 | 971 | 73 | ||
| AK133756.1 | - | 9 - 10 | 974 | 73 | ||
| AK154269.1 | - | 9 - 10 | 974 | 73 | ||
| AK170658.1 | - | 9 - 10 | 978 | 73 | ||
| BC013495.1 | - | 9 - 10 | 966 | 73 | ||
| BC023343.1 | - | 9 - 10 | 960 | 5136781 | 73 | |
| X57199.1 | - | 9 - 10 | 949 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acp2
Genomic Map