Assay Details
| Gene Symbol: Lnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110209.1 | NP_001103679.1 | 1 - 2 | 134 | 110 | |
| NM_027133.3 | NP_081409.1 | 1 - 2 | 134 | 110 | ||
| GenBank mRNA | AK009311.1 | - | 1 - 2 | 108 | 110 | |
| AK053593.1 | - | 1 - 2 | 103 | 110 | ||
| AK082900.1 | - | 1 - 2 | 118 | 110 | ||
| AK136506.1 | - | 1 - 2 | 110 | 109 | ||
| AK137445.1 | - | 1 - 2 | 135 | 110 | ||
| AK143497.1 | - | 1 - 2 | 103 | 110 | ||
| AK173241.1 | - | 1 - 2 | 66 | 110 | ||
| AY312281.1 | - | 1 - 2 | 55 | 110 | ||
| BC057961.1 | - | 1 - 2 | 61 | 5353629 | 110 | |
| BY161702.1 | - | 1 - 2 | 130 | 110 | ||
| BY770278.1 | - | 1 - 2 | 134 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK136506.1, AK143497.1, AY312281.1, BC057961.1, NM_001110209.1, NM_027133.3 transcripts.
Set Membership: |
|

Lnp
Genomic Map