Assay Details
| Gene Symbol: Lnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110209.1 | NP_001103679.1 | 7 - 8 | 631 | 90 | |
| NM_027133.3 | NP_081409.1 | 7 - 8 | 631 | 90 | ||
| GenBank mRNA | AK014897.1 | - | 7 - 8 | 529 | 90 | |
| AK136506.1 | - | 7 - 8 | 606 | 90 | ||
| AK137445.1 | - | 7 - 8 | 632 | 90 | ||
| AK173241.1 | - | 7 - 8 | 563 | 90 | ||
| AY312281.1 | - | 7 - 8 | 552 | 90 | ||
| BC057961.1 | - | 7 - 8 | 558 | 5353629 | 90 | |
| BC060153.1 | - | 6 - 7 | 470 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lnp
Genomic Map