Assay Details
| Gene Symbol: Cpa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027926.2 | NP_082202.1 | 5 - 6 | 501 | 60 | |
| GenBank mRNA | AK030705.1 | - | 5 - 6 | 501 | 60 | |
| BC061206.1 | - | 5 - 6 | 508 | 30249166 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpa4
Genomic Map