Assay Details
| Gene Symbol: Ocm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033039.3 | NP_149028.2 | 2 - 3 | 195 | 82 | |
| GenBank mRNA | BQ567296.1 | - | 2 - 3 | 84 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2073825 | NM_033039.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094916 | NM_033039.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094916 | BQ567296.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297705 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616540 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 831644 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375917 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 805488 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 794465 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966101 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 480239 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236085 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 182188 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952538 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815397 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40514 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40254 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 598935 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917004 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 102622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 42967 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 56284 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122182 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2073825 | NM_033039.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094916 | NM_033039.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094916 | BQ567296.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159203 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01242112 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05776415 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122182 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05776378 | - | - |
| Whole Mouse Genome G4122A | A_52_P79648 | NM_033039.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P79648 | BQ567296.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100173 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104469 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05776415 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04148843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04859964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04425990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04511155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04552529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04121272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04509943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04552532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04509946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04280649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04868256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04730091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04575610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04956041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04102738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04730090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04413885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04206745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04121276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04609612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04620615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04524688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04859965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04818018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04623606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159223 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10535529 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535530 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535527 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535526 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535531 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535525 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535528 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10535524 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500032730.mm.1 | NM_033039.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032730.mm.1 | NM_033039.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032730.mm.1 | TC0500003569.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032735.mm.1 | NM_033039.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032735.mm.1 | NM_033039.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032735.mm.1 | TC0500003569.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032729.mm.1 | NM_033039.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032729.mm.1 | NM_033039.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032729.mm.1 | TC0500003569.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 101174_at | - | - |
| Murine Genome U74v2 Set | 101174_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 447794 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4480722 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480722 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1233351 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1233351 | - | - |
| mouse-6 v1.1 (Illumina) | scl25796.6.1_22-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480722 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25796.6.1_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480722 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1233351 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1233351 | - | - |

Ocm
Genomic Map