Assay Details
| Gene Symbol: Bsdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133889.2 | NP_598650.2 | 6 - 7 | 546 | 64 | |
| GenBank mRNA | AK037201.1 | - | 6 - 7 | 542 | 64 | |
| AK144053.1 | - | 6 - 7 | 541 | 64 | ||
| BC021480.1 | - | 5 - 6 | 526 | 64 | ||
| BC049111.1 | - | 6 - 7 | 546 | 6415574 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bsdc1
Genomic Map