Assay Details
| Gene Symbol: Scaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134123.3 | NP_598884.2 | 4 - 5 | 735 | 153 | |
| GenBank mRNA | AK122451.1 | - | 4 - 5 | 602 | 153 | |
| AK171078.1 | - | 5 - 6 | 542 | 153 | ||
| BC038363.1 | - | 4 - 5 | 379 | 5371946 | 153 | |
| BC075621.1 | - | 4 - 5 | 728 | 6406441 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scaf8
Genomic Map