Assay Details
| Gene Symbol: Cobll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027225.1 | NP_081501.1 | 7 - 8 | 1362 | 70 | |
| NM_177025.5 | NP_795999.2 | 7 - 8 | 1362 | 70 | ||
| GenBank mRNA | AK037010.1 | - | 8 - 9 | 1353 | 70 | |
| AK052533.1 | - | 7 - 8 | 1311 | 70 | ||
| AK144943.1 | - | 8 - 9 | 1406 | 70 | ||
| AK164624.1 | - | 8 - 9 | 1292 | 70 | ||
| AK173076.1 | - | 7 - 8 | 1193 | 70 | ||
| AY308746.1 | - | 7 - 8 | 1362 | 70 | ||
| BC067007.1 | - | 7 - 8 | 1359 | 5702930 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cobll1
Genomic Map