Assay Details
| Gene Symbol: Cobll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177025.5 | NP_795999.2 | 8 - 9 | 1443 | 69 | |
| GenBank mRNA | AK037010.1 | - | 9 - 10 | 1434 | 69 | |
| AK144943.1 | - | 9 - 10 | 1487 | 69 | ||
| AY308746.1 | - | 8 - 9 | 1443 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cobll1
Genomic Map