Assay Details
| Gene Symbol: Ccz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177682.3 | NP_808350.1 | 4 - 5 | 427 | 98 | |
| GenBank mRNA | AK090008.1 | - | 4 - 5 | 441 | 98 | |
| AK146704.1 | - | 4 - 5 | 460 | 98 | ||
| AK150793.1 | - | 4 - 5 | 464 | 98 | ||
| AK152328.1 | - | 4 - 5 | 463 | 98 | ||
| AK152576.1 | - | 4 - 5 | 463 | 98 | ||
| AK153160.1 | - | 4 - 5 | 464 | 98 | ||
| BC048077.1 | - | 4 - 5 | 427 | 6308253 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccz1
Genomic Map