Assay Details
| Gene Symbol: Ccz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177682.3 | NP_808350.1 | 8 - 9 | 821 | 106 | |
| GenBank mRNA | AK090008.1 | - | 8 - 9 | 835 | 106 | |
| AK150793.1 | - | 8 - 9 | 858 | 106 | ||
| AK152328.1 | - | 8 - 9 | 857 | 106 | ||
| AK152576.1 | - | 8 - 9 | 857 | 106 | ||
| AK153160.1 | - | 8 - 9 | 858 | 106 | ||
| BC048077.1 | - | 8 - 9 | 821 | 6308253 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24159038 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24159040 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24159037 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24159039 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395293 | BC048077.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395293 | AK216274.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395293 | AK146704.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395293 | AK090008.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395293 | NM_177682.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395293 | BC048077.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395293 | AK216274.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395293 | AK146704.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395293 | AK090008.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395293 | NM_177682.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481519 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 269252 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316833 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237186 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142489 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729292 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 742447 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 691617 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 479852 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406911 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 421665 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469359 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545994 | M38129.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545994 | M74753.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545994 | NM_001099635.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545994 | XM_908146.3 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 315859 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874202 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17024 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 178460 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143724 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118893 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395293 | BC048077.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395293 | AK216274.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395293 | AK146704.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395293 | AK090008.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395293 | NM_177682.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395293 | BC048077.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395293 | AK216274.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395293 | AK146704.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395293 | AK090008.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395293 | NM_177682.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24159056 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05776325 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05776315 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118893 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143724 | - | - |
| Whole Mouse Genome G4122A | A_51_P395293 | BC048077.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395293 | AK216274.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395293 | AK146704.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395293 | AK090008.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395293 | NM_177682.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395293 | BC048077.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395293 | AK216274.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395293 | AK146704.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395293 | AK090008.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395293 | NM_177682.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105468 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143724 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118893 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04151948 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24159038 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24159040 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24159039 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24159037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24159023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04367295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04520268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04767683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04393227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04367292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04814550 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04151948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04114382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04814549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04871320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04820781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04814551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04737726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04114381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04114378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04220518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04437697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24159029 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10535510 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535522 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535517 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535520 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535513 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535516 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535518 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535515 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535519 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535512 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535521 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535511 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535514 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535523 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535509 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433554_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10535508 | - | - |
| Mouse Genome 430 2.0 Array | 1433554_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433554_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231874_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231874_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500032715.mm.1 | NM_177682.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032715.mm.1 | NM_177682.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032715.mm.1 | TC0500003568.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430A Array | 1433554_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433554_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 352634 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3170332 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3170332 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003170332 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2767674 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2767674 | - | - |
| mouse-6 v1.1 (Illumina) | scl25797.15.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3170332 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25797.15.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3170332 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2767674 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2767674 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl25797.15.1_2-S | - | - |

Ccz1
Genomic Map