Assay Details
| Gene Symbol: Klhl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177771.5 | NP_808439.2 | 3 - 4 | 504 | 57 | |
| GenBank mRNA | AK044774.1 | - | 3 - 4 | 615 | 57 | |
| AK173029.1 | - | 3 - 4 | 504 | 57 | ||
| BC025563.1 | - | 3 - 4 | 500 | 5322893 | 57 | |
| BY248410.1 | - | 3 - 4 | 424 | 57 | ||
| CK619719.1 | - | 1 - 2 | 110 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC025563.1 transcripts.
Set Membership: |
|

Klhl18
Genomic Map