Assay Details
| Gene Symbol: Mief1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178719.5 | NP_848834.2 | 4 - 5 | 774 | 76 | |
| GenBank mRNA | AK038466.1 | - | 4 - 5 | 747 | 76 | |
| AK044773.1 | - | 5 - 6 | 1207 | 76 | ||
| AK050546.1 | - | 4 - 5 | 735 | 76 | ||
| AK080375.1 | - | 4 - 5 | 720 | 76 | ||
| AK163389.1 | - | 5 - 6 | 875 | 76 | ||
| BC094597.1 | - | 4 - 5 | 706 | 76 | ||
| BC137633.1 | - | 3 - 4 | 678 | 8860652 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mief1
Genomic Map