Assay Details
| Gene Symbol: Zpld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178720.4 | NP_848835.1 | 10 - 11 | 1200 | 65 | |
| GenBank mRNA | AK034628.1 | - | 10 - 11 | 1201 | 65 | |
| AK047981.1 | - | 9 - 10 | 1110 | 65 | ||
| AK052599.1 | - | 11 - 12 | 1310 | 65 | ||
| BC116255.1 | - | 10 - 11 | 1169 | 40093632 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zpld1
Genomic Map