Assay Details
| Gene Symbol: Zpld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178720.4 | NP_848835.1 | 2 - 3 | 151 | 65 | |
| GenBank mRNA | AK034628.1 | - | 2 - 3 | 152 | 65 | |
| AK047981.1 | - | 1 - 2 | 61 | 65 | ||
| AK052599.1 | - | 3 - 4 | 261 | 65 | ||
| BC116254.1 | - | 2 - 3 | 120 | 40093631 | 65 | |
| BC116255.1 | - | 2 - 3 | 120 | 40093632 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK034628.1, AK047981.1, AK052599.1, BC116254.1, BC116255.1, NM_178720.4 transcripts.
Set Membership: |
|

Zpld1
Genomic Map