| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000020168.mm.1 |
NM_001277188.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020168.mm.1 |
NR_102339.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020168.mm.1 |
TC1000002653.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000020166.mm.1 |
NM_001277188.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020166.mm.1 |
NR_102339.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020166.mm.1 |
TC1000002653.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000020169.mm.1 |
NM_001277188.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020169.mm.1 |
NR_102339.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020169.mm.1 |
TC1000002653.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000020167.mm.1 |
NM_001277188.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020167.mm.1 |
NR_102339.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000020167.mm.1 |
TC1000002653.mm.1 |
MTA TC |