Assay Details
| Gene Symbol: Ano4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277188.1 | NP_001264117.1 | 7 - 8 | 1100 | 58 | |
| NR_102339.1 | 6 - 7 | 1057 | 58 | |||
| GenBank mRNA | AK039466.1 | - | 7 - 8 | 964 | 58 | |
| AK039728.1 | - | 7 - 8 | 910 | 58 | ||
| AK078364.1 | - | 7 - 8 | 864 | 58 | ||
| BC065114.1 | - | 6 - 7 | 1057 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ano4
Genomic Map