Assay Details
| Gene Symbol: Tmem88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025915.4 | NP_080191.3 | 2 | 1055 | 69 | |
| GenBank mRNA | AK045149.1 | - | 2 | 1056 | 69 | |
| AK161789.1 | - | 2 | 1056 | 69 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209484 | AK161789.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209484 | BC100316.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209484 | NM_025915.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209484 | AK010789.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209484 | AK011240.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209484 | AK161789.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209484 | BC100316.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209484 | NM_025915.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209484 | AK010789.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209484 | AK011240.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209484 | AK161789.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209484 | BC100316.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209484 | NM_025915.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209484 | AK010789.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209484 | AK011240.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209484 | AK161789.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209484 | BC100316.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209484 | NM_025915.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209484 | AK010789.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209484 | AK011240.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894834 | - | - |
| Whole Mouse Genome G4122A | A_52_P209484 | AK161789.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209484 | BC100316.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209484 | NM_025915.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209484 | AK010789.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209484 | AK011240.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209484 | AK161789.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209484 | BC100316.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209484 | NM_025915.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209484 | AK010789.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209484 | AK011240.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135557 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09628621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09694615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09628611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09628620 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10387370 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387371 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377546 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387369 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453285_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10387368 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10377541 | - | - |
| Mouse Genome 430 2.0 Array | 1453285_at | - | - |
| Mouse Genome 430 2.0 Array | 1453285_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67020_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67020_at | - | - |
| Mouse Expression Set 430 | 1453285_at | - | - |
| Mouse Expression Set 430 | 1453285_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453285_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453285_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 338672 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 520204 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 520204 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000520204 | - | - |
| mouse-6 v1.1 (Illumina) | scl40036.2_49-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 520204 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40036.2_49-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 520204 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2822321 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2822321 | - | - |

Tmem88
Genomic Map