Assay Details
| Gene Symbol: Fam32a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026455.4 | NP_080731.1 | 4 | 1344 | 114 | |
| GenBank mRNA | AB041658.1 | - | 4 | 1342 | 114 | |
| AK007524.1 | - | 4 | 1431 | 114 | ||
| AK011922.1 | - | 4 | 1330 | 114 | ||
| AK075976.1 | - | 4 | 1345 | 114 | ||
| AK082792.1 | - | 4 | 1315 | 114 | ||
| BC006058.1 | - | 4 | 1296 | 3591395 | 114 | |
| BC085487.1 | - | 3 | 3155 | 114 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25960242 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25960244 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25960243 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549977 | BC006058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549977 | BC085487.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549977 | AK011922.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549977 | AK075976.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549977 | AK082792.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549977 | BC006058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549977 | BC085487.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549977 | AK011922.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549977 | AK075976.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549977 | AK082792.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P482124 | BC006058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P482124 | BC085487.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P482124 | AK011922.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P482124 | AK075976.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P482124 | AK082792.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P482124 | BC006058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P482124 | BC085487.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P482124 | AK011922.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P482124 | AK075976.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P482124 | AK082792.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 629301 | AK085379.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355630 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 247760 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 3724 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135495 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P482124 | BC006058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P482124 | BC085487.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P482124 | AK011922.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P482124 | AK075976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P482124 | AK082792.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P482124 | BC006058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P482124 | BC085487.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P482124 | AK011922.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P482124 | AK075976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P482124 | AK082792.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549977 | BC006058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549977 | BC085487.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549977 | AK011922.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549977 | AK075976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549977 | AK082792.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549977 | BC006058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549977 | BC085487.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549977 | AK011922.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549977 | AK075976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549977 | AK082792.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960243 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135495 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06552751 | - | - |
| Whole Mouse Genome G4122A | A_52_P549977 | BC006058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P549977 | BC085487.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P549977 | AK011922.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P549977 | AK075976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P549977 | AK082792.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P549977 | BC006058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P549977 | BC085487.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P549977 | AK011922.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P549977 | AK075976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P549977 | AK082792.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482124 | BC006058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482124 | BC085487.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482124 | AK011922.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482124 | AK075976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482124 | AK082792.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482124 | BC006058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482124 | BC085487.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482124 | AK011922.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482124 | AK075976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482124 | AK082792.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107473 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135495 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07216814 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07216810 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25960242 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25960244 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25960243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07228723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07373466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06772761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07216816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07073848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06976773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07073849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06976777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960247 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10572784 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572785 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572782 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572780 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572783 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562600 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572781 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423217_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423216_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10562599 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10572779 | - | - |
| Mouse Genome 430 2.0 Array | 1423216_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423217_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423216_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423217_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67922_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67922_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800006014.mm.1 | NM_026455.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006014.mm.1 | NM_026455.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006014.mm.1 | TC0800000818.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800006015.mm.1 | NM_026455.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006015.mm.1 | NM_026455.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006015.mm.1 | TC0800000818.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800006017.mm.1 | NM_026455.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006017.mm.1 | NM_026455.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006017.mm.1 | TC0800000818.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800006016.mm.1 | NM_026455.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006016.mm.1 | NM_026455.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006016.mm.1 | TC0800000818.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800006019.mm.1 | NM_026455.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006019.mm.1 | NM_026455.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006019.mm.1 | TC0800000818.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800006020.mm.1 | NM_026455.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006020.mm.1 | NM_026455.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006020.mm.1 | TC0800000818.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423216_a_at | - | - |
| Mouse Expression Set 430 | 1423216_PM_a_at | - | - |
| Mouse Expression Set 430 | 1423217_PM_a_at | - | - |
| Mouse Expression Set 430 | 1423217_a_at | - | - |
| Murine Genome U74v2 Set | 97866_PM_at | - | - |
| Murine Genome U74v2 Set | 97865_PM_g_at | - | - |
| Murine Genome U74v2 Set | 97866_at | - | - |
| Murine Genome U74v2 Set | 97864_at | - | - |
| Murine Genome U74v2 Set | 97864_PM_at | - | - |
| Murine Genome U74v2 Set | 97865_g_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423216_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423217_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423217_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423216_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423217_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423217_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423216_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423216_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 862922 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3780048 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003780048 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223730 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223730 | - | - |
| mouse-6 v1.1 (Illumina) | scl067922.4_27-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3780048 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067922.4_27-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3780048 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2778799 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2778799 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223730 | - | - |

Fam32a
Genomic Map