Assay Details
| Gene Symbol: Olfr123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146630.1 | NP_666841.1 | 1 | 850 | 84 | |
| GenBank mRNA | BC148279.1 | - | 1 | 850 | 40136837 | 84 |
| BC148324.1 | - | 1 | 850 | 40136838 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P125253 | NM_146630.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P125253 | BC148279.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P125253 | BC148324.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P125253 | NM_146630.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P125253 | BC148279.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P125253 | BC148324.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125785 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P125253 | NM_146630.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P125253 | BC148279.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P125253 | BC148324.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P125253 | NM_146630.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P125253 | BC148279.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P125253 | BC148324.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P125253 | NM_146630.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P125253 | BC148279.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P125253 | BC148324.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P125253 | NM_146630.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P125253 | BC148279.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P125253 | BC148324.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169934 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125785 | - | - |
| Whole Mouse Genome G4122A | A_52_P125253 | NM_146630.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P125253 | BC148279.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P125253 | BC148324.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P125253 | NM_146630.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P125253 | BC148279.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P125253 | BC148324.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109408 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125785 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13378939 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13378918 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13378949 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13378943 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13378957 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13378938 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13378930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13378917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13378922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13093676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13078821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31169926 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10445157 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10445156 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 538399 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2030070 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2030070 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002030070 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2691011 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2691011 | - | - |
| mouse-6 v1.1 (Illumina) | scl8513.1.1_62-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2030070 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl8513.1.1_62-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2030070 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2900151 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2900151 | - | - |

Olfr123
Genomic Map