Assay Details
| Gene Symbol: Olfr1428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146678.2 | NP_666889.2 | 1 | 908 | 72 | |
| GenBank mRNA | BC119441.2 | - | 1 | 908 | 40042424 | 72 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054847 | NM_146678.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054847 | BC119441.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61407 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104304 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054847 | NM_146678.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054847 | BC119441.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P163713 | NM_146678.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P163713 | BC119441.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959541 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104304 | - | - |
| Whole Mouse Genome G4122A | A_51_P163713 | NM_146678.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163713 | BC119441.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133654 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104304 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14285206 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14285223 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14285207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14285206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959543 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14246020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31959595 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10466289 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10466288 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900010772.mm.1 | NM_146678.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010772.mm.1 | NM_146678.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010772.mm.1 | TC1900001155.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 533546 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3450520 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3450520 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003450520 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244582 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244582 | - | - |
| mouse-6 v1.1 (Illumina) | scl52711.1.1_41-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3450520 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52711.1.1_41-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3450520 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1244582 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244582 | - | - |

Olfr1428
Genomic Map