Assay Details
| Gene Symbol: Nnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291128.1 | NP_001278057.1 | 3 | 482 | 73 | |
| NM_001291129.1 | NP_001278058.1 | 3 | 470 | 73 | ||
| NM_001291130.1 | NP_001278059.1 | 3 | 421 | 73 | ||
| NM_010923.3 | NP_035053.1 | 3 | 475 | 73 | ||
| NM_180960.3 | NP_851291.1 | 2 | 394 | 73 | ||
| GenBank mRNA | AK160626.1 | - | 3 | 443 | 73 | |
| AU079843.1 | - | 2 | 361 | 73 | ||
| AY644389.1 | - | 1 | 204 | 73 | ||
| BC036984.1 | - | 3 | 433 | 4981517 | 73 | |
| GQ184462.1 | - | 3 | 376 | 73 | ||
| X83568.1 | - | 2 | 292 | 73 | ||
| X83569.1 | - | 2 | 349 | 73 | ||
| X83570.1 | - | 3 | 430 | 73 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21777849 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21777852 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21777847 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21777850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21777851 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21777848 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21777853 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980426 | X83569.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980426 | X83570.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980426 | X83568.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980426 | AK160626.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980426 | AY644389.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P484956 | X83569.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P484956 | X83570.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P484956 | X83568.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P484956 | AK160626.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P484956 | AK003004.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P484956 | X83569.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P484956 | X83570.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P484956 | X83568.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P484956 | AK160626.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P484956 | AK003004.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P484956 | X83569.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P484956 | X83570.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P484956 | X83568.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P484956 | AK160626.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P484956 | AK003004.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P484956 | X83569.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P484956 | X83570.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P484956 | X83568.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P484956 | AK160626.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P484956 | AK003004.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980426 | X83569.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980426 | X83570.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980426 | X83568.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980426 | AK160626.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980426 | AY644389.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21777856 | - | - |
| Whole Mouse Genome G4122A | A_52_P484956 | X83569.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P484956 | X83570.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P484956 | X83568.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P484956 | AK160626.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P484956 | AK003004.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P484956 | X83569.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P484956 | X83570.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P484956 | X83568.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P484956 | AK160626.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P484956 | AK003004.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100474 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01574991 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01574988 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01574990 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01574983 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01574982 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01574979 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21777852 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21777847 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21777848 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21777849 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21777851 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21777850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21777853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21777882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01574988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01693274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01574977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01574991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21777823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21777837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01225152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01743580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21777844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21777840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01693273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21777843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21777853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21777855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01225154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01528455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21777822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01483776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21777856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21777859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21777839 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10477987 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477989 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477990 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477988 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423506_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10477986 | - | - |
| Mouse Genome 430 2.0 Array | 1423506_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423506_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18111_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18111_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200019558.mm.1 | NM_010923.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019558.mm.1 | NM_180960.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019558.mm.1 | TC0200002411.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200019560.mm.1 | NM_010923.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019560.mm.1 | NM_180960.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019560.mm.1 | TC0200002411.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200019556.mm.1 | NM_010923.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019556.mm.1 | NM_180960.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019556.mm.1 | TC0200002411.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200019557.mm.1 | NM_010923.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019557.mm.1 | NM_180960.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019557.mm.1 | TC0200002411.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200019559.mm.1 | NM_010923.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019559.mm.1 | NM_180960.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200019559.mm.1 | TC0200002411.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423506_PM_a_at | - | - |
| Mouse Expression Set 430 | 1423506_a_at | - | - |
| Murine Genome U74v2 Set | 97520_s_at | - | - |
| Murine Genome U74v2 Set | 97520_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423506_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423506_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423506_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423506_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100430487 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5130253 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4920053 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100430487 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5130253 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004920053 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005130253 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232456 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2755578 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2755578 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232456 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237036 | - | - |
| mouse-6 v1.1 (Illumina) | scl0003133.1_10-S | - | - |
| mouse-6 v1.1 (Illumina) | scl20011.1_69-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0018111.2_84-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4920053 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5130253 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100430487 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003133.1_10-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0018111.2_84-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100430487 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4920053 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5130253 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2755578 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1232456 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232456 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237036 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2755578 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003133.1_10-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0018111.2_84-S | - | - |

Nnat
Genomic Map