Assay Details
| Gene Symbol: Cept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133869.4 | NP_598630.2 | 5 - 6 | 913 | 85 | |
| GenBank mRNA | AK037116.1 | - | 5 - 6 | 873 | 85 | |
| BC023783.1 | - | 5 - 6 | 853 | 5346896 | 85 | |
More Information
Set Membership: |
|

Cept1
Genomic Map