Assay Details
| Gene Symbol: Dusp28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175118.3 | NP_780327.1 | 1 - 2 | 435 | 137 | |
| GenBank mRNA | AK002934.1 | - | 1 - 2 | 434 | 137 | |
| AK088940.1 | - | 1 - 2 | 382 | 137 | ||
| AK163560.1 | - | 1 - 2 | 444 | 137 | ||
| BC051598.1 | - | 1 - 2 | 412 | 137 | ||
| BC119127.1 | - | 1 - 2 | 424 | 8733876 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20451937 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20451934 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20451933 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20451932 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20451935 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249793 | AW493803.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249793 | NM_175118.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249793 | AK002934.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249793 | AK088940.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249793 | AK163560.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249793 | AW493803.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249793 | NM_175118.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249793 | AK002934.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249793 | AK088940.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249793 | AK163560.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109137 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249793 | AW493803.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249793 | NM_175118.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249793 | AK002934.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249793 | AK088940.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249793 | AK163560.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249793 | AW493803.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249793 | NM_175118.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249793 | AK002934.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249793 | AK088940.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249793 | AK163560.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20451934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20451942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20451946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20451937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20451945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20451932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20451944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20451940 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109137 | - | - |
| Whole Mouse Genome G4122A | A_51_P249793 | AW493803.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249793 | NM_175118.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249793 | AK002934.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249793 | AK088940.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249793 | AK163560.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249793 | AW493803.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249793 | NM_175118.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249793 | AK002934.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249793 | AK088940.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249793 | AK163560.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138188 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109137 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00919169 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20451932 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20451933 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20451934 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20451935 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20451937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20452009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20451899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20451945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20451922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00273457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20451926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00753683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20451934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00919175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20451928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20451940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00251686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00251688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20451942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20451927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20451917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20451937 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10348669 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348667 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10348668 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429613_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10348666 | - | - |
| Mouse Genome 430 2.0 Array | 1429613_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429613_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67446_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67446_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100007697.mm.1 | NM_175118.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100007697.mm.1 | NM_175118.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100007697.mm.1 | TC0100000919.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100007698.mm.1 | NM_175118.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100007698.mm.1 | NM_175118.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100007698.mm.1 | TC0100000919.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100007696.mm.1 | NM_175118.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100007696.mm.1 | NM_175118.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100007696.mm.1 | TC0100000919.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429613_at | - | - |
| Mouse Expression Set 430 | 1429613_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429613_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429613_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 586228 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4480088 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480088 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2776681 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2776681 | - | - |
| mouse-6 v1.1 (Illumina) | scl17636.2.1_21-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480088 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl17636.2.1_21-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480088 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2776681 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2776681 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl17636.2.1_21-S | - | - |

Dusp28
Genomic Map