Assay Details
| Gene Symbol: 4933421I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027702.2 | NP_081978.1 | 3 - 4 | 398 | 144 | |
| GenBank mRNA | AK016864.1 | - | 3 - 4 | 399 | 144 | |
| BC147587.1 | - | 3 - 4 | 438 | 9056577 | 144 | |
| BC147628.1 | - | 3 - 4 | 438 | 9056618 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275268 | AK016864.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275268 | NM_027702.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275268 | AK016864.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275268 | NM_027702.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 384876 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 153832 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168682 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275268 | AK016864.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275268 | NM_027702.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275268 | AK016864.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275268 | NM_027702.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25079714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25079704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25079700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25079729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25079746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25079737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25079708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25079701 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168682 | - | - |
| Whole Mouse Genome G4122A | A_51_P275268 | AK016864.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275268 | NM_027702.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275268 | AK016864.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275268 | NM_027702.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124209 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168682 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06221445 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06333748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25079690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25079750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25079757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25079678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25079722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25079684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25079711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06333729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06293779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06519433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06113012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06023197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05983292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06333748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06333734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06333751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06333730 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10562645 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562644 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562646 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449995_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10562643 | - | - |
| Mouse Genome 430 2.0 Array | 1449995_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449995_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71162_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71162_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700025975.mm.1 | NM_027702.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025975.mm.1 | NM_027702.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025975.mm.1 | TC0700002922.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025976.mm.1 | NM_027702.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025976.mm.1 | NM_027702.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025976.mm.1 | TC0700002922.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025974.mm.1 | NM_027702.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025974.mm.1 | NM_027702.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025974.mm.1 | TC0700002922.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025978.mm.1 | TC0700002922.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025977.mm.1 | NM_027702.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025977.mm.1 | NM_027702.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025977.mm.1 | TC0700002922.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025973.mm.1 | NM_027702.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025973.mm.1 | NM_027702.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025973.mm.1 | TC0700002922.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449995_at | - | - |
| Mouse Expression Set 430 | 1449995_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449995_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449995_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449995_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449995_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 769631 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3800484 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003800484 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255826 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255826 | - | - |
| mouse-6 v1.1 (Illumina) | scl31432.4.1_268-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3800484 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31432.4.1_268-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3800484 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1255826 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255826 | - | - |

4933421I07Rik
Genomic Map