Assay Details
| Gene Symbol: Vmn1r54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053224.1 | NP_444454.1 | 1 | 842 | 149 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441585 | NM_053224.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441585 | NM_053224.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342000 | AK134012.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342000 | AK154070.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342000 | BC013274.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342000 | NM_011488.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342000 | U21103.1 | genbank_mrna_mouse |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136460 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441585 | NM_053224.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441585 | NM_053224.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P441585 | NM_053224.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P441585 | NM_053224.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24631412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24631409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24631424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24631415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24631433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24631427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24631426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24631416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24631425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24631429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24631413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24631411 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136460 | - | - |
| Whole Mouse Genome G4122A | A_51_P441585 | NM_053224.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P441585 | NM_053224.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129213 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136460 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05576213 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05576229 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05576217 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05576223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05294467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05576217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05576223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05865162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05576220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24631434 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10539982 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422366_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539981 | - | - |
| Mouse Genome 430 2.0 Array | 1422366_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422366_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 113851_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 113851_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600008271.mm.1 | NM_053224.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600008271.mm.1 | NM_053224.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600008271.mm.1 | TC0600001068.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422366_at | - | - |
| Mouse Expression Set 430 | 1422366_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422366_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422366_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422366_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422366_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 468597 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6900465 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006900465 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2429488 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2429488 | - | - |
| mouse-6 v1.1 (Illumina) | scl29757.1.1_330-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6900465 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29757.1.1_330-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900465 | - | - |

Vmn1r54
Genomic Map