Assay Details
| Gene Symbol: Stoml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023231.2 | NP_075720.1 | 10 | 1449 | 140 | |
| GenBank mRNA | AK002793.1 | - | 10 | 1446 | 140 | |
| BC003425.1 | - | 10 | 1449 | 2609584 | 140 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial calcium ion transport
cellular calcium ion homeostasis
mitochondrion organization
lipid localization
positive regulation of mitochondrial membrane potential
interleukin-2 production
mitochondrial protein processing
CD4-positive, alpha-beta T cell activation
mitochondrial ATP synthesis coupled proton transport
T cell receptor signaling pathway
protein oligomerization
mitochondrion localization
positive regulation of mitochondrial DNA replication
positive regulation of cardiolipin metabolic process
stress-induced mitochondrial fusion
lipid binding
GTPase binding
cardiolipin binding
mitochondrial calcium ion transport
cellular calcium ion homeostasis
mitochondrion organization
lipid localization
positive regulation of mitochondrial membrane potential
interleukin-2 production
mitochondrial protein processing
CD4-positive, alpha-beta T cell activation
mitochondrial ATP synthesis coupled proton transport
T cell receptor signaling pathway
protein oligomerization
mitochondrion localization
positive regulation of mitochondrial DNA replication
positive regulation of cardiolipin metabolic process
stress-induced mitochondrial fusion
lipid binding
GTPase binding
cardiolipin binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22869602 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22869603 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22869599 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22869604 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958808 | BC003425.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958808 | BC069941.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958808 | NM_023231.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958808 | AF323178.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958808 | AK002793.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028922 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 912650 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 110951 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16115 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156494 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028922 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958808 | BC003425.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958808 | BC069941.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958808 | NM_023231.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958808 | AF323178.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958808 | AK002793.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869593 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869601 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22869605 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156494 | - | - |
| Whole Mouse Genome G4122A | A_52_P187787 | BC003425.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P187787 | BC069941.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P187787 | NM_023231.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P187787 | AF323178.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P187787 | AK002793.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380685 | BC003425.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380685 | BC069941.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380685 | NM_023231.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380685 | AF323178.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380685 | AK002793.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120037 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110370 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114980 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156494 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03256535 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03256559 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03902105 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03256570 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03256547 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03256548 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22869602 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22869604 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22869599 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22869603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22869577 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22869605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22869596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03256569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22869611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03256060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22869595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22869608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03256556 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03256570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22869609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22869578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22869582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03256549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03256572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22869603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22869587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22869599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22869576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03256064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03256566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03256559 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10512446 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512447 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406204 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512448 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512444 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512450 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512445 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512449 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448774_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10406203 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10512443 | - | - |
| Mouse Genome 430 2.0 Array | 1448774_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448774_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66592_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66592_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400020386.mm.1 | NM_023231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020386.mm.1 | NM_023231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020386.mm.1 | TC0400002516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020389.mm.1 | NM_023231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020389.mm.1 | NM_023231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020389.mm.1 | TC0400002516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020395.mm.1 | NM_023231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020395.mm.1 | NM_023231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020395.mm.1 | TC0400002516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020396.mm.1 | NM_023231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020396.mm.1 | NM_023231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020396.mm.1 | TC0400002516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020380.mm.1 | NM_023231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020380.mm.1 | NM_023231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020380.mm.1 | TC0400002516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020385.mm.1 | NM_023231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020385.mm.1 | NM_023231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020385.mm.1 | TC0400002516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020409.mm.1 | NM_023231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020409.mm.1 | NM_023231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020409.mm.1 | TC0400002516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020403.mm.1 | NM_023231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020403.mm.1 | NM_023231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020403.mm.1 | TC0400002516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020388.mm.1 | NM_023231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020388.mm.1 | NM_023231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020388.mm.1 | TC0400002516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020383.mm.1 | NM_023231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020383.mm.1 | NM_023231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020383.mm.1 | TC0400002516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020405.mm.1 | NM_023231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020405.mm.1 | NM_023231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020405.mm.1 | TC0400002516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020400.mm.1 | NM_023231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020400.mm.1 | NM_023231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020400.mm.1 | TC0400002516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020391.mm.1 | NM_023231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020391.mm.1 | NM_023231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020391.mm.1 | TC0400002516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020401.mm.1 | NM_023231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020401.mm.1 | NM_023231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020401.mm.1 | TC0400002516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020412.mm.1 | NM_023231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020412.mm.1 | NM_023231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020412.mm.1 | TC0400002516.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448774_PM_at | - | - |
| Mouse Expression Set 430 | 1448774_at | - | - |
| Murine Genome U74v2 Set | 96289_PM_at | - | - |
| Murine Genome U74v2 Set | 96289_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448774_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448774_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448774_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448774_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 641879 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1940487 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1940487 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001940487 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2721799 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2721799 | - | - |
| mouse-6 v1.1 (Illumina) | scl24408.10_6-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1940487 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24408.10_6-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1940487 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2721799 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2721799 | - | - |

Stoml2
Genomic Map