Assay Details
| Gene Symbol: Hpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008277.2 | NP_032303.1 | 10 - 11 | 842 | 55 | |
| GenBank mRNA | AK149416.1 | - | 11 - 12 | 880 | 55 | |
| BC013343.1 | - | 10 - 11 | 866 | 4193670 | 55 | |
| D29987.1 | - | 11 - 12 | 880 | 55 | ||
| X59530.1 | - | 10 - 11 | 832 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hpd
Genomic Map