Assay Details
| Gene Symbol: Abl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112703.2 | NP_001106174.1 | 8 - 9 | 2754 | 62 | |
| NM_001283045.1 | NP_001269974.1 | 8 - 9 | 1631 | 62 | ||
| NM_001283046.1 | NP_001269975.1 | 9 - 10 | 2959 | 62 | ||
| NM_001283047.1 | NP_001269976.1 | 9 - 10 | 2898 | 62 | ||
| NM_009594.4 | NP_033724.2 | 8 - 9 | 1525 | 62 | ||
| GenBank mRNA | AK088252.1 | - | 9 - 10 | 2884 | 62 | |
| AK090095.1 | - | 8 - 9 | 1483 | 62 | ||
| AK166028.1 | - | 8 - 9 | 2743 | 62 | ||
| AK166210.1 | - | 9 - 10 | 2993 | 62 | ||
| BC059260.1 | - | 8 - 9 | 1552 | 6408990 | 62 | |
| BC103770.1 | - | 8 - 9 | 1978 | 30848553 | 62 | |
| CF726668.1 | - | 2 - 3 | 238 | 62 | ||
| J02995.1 | - | 8 - 9 | 1478 | 62 | ||
| L10656.1 | - | 6 - 7 | 1079 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abl1
Genomic Map