Assay Details
| Gene Symbol: Abl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112703.2 | NP_001106174.1 | 9 - 10 | 2839 | 63 | |
| NM_001283045.1 | NP_001269974.1 | 9 - 10 | 1716 | 63 | ||
| NM_001283046.1 | NP_001269975.1 | 10 - 11 | 3044 | 63 | ||
| NM_001283047.1 | NP_001269976.1 | 10 - 11 | 2983 | 63 | ||
| NM_009594.4 | NP_033724.2 | 9 - 10 | 1610 | 63 | ||
| GenBank mRNA | AK088252.1 | - | 10 - 11 | 2969 | 63 | |
| AK090095.1 | - | 9 - 10 | 1568 | 63 | ||
| AK166028.1 | - | 9 - 10 | 2828 | 63 | ||
| AK166210.1 | - | 10 - 11 | 3078 | 63 | ||
| BC059260.1 | - | 9 - 10 | 1637 | 6408990 | 63 | |
| BC103770.1 | - | 9 - 10 | 2063 | 30848553 | 63 | |
| CF726668.1 | - | 3 - 4 | 323 | 63 | ||
| J02995.1 | - | 9 - 10 | 1563 | 63 | ||
| L10656.1 | - | 7 - 8 | 1164 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abl1
Genomic Map