Assay Details
| Gene Symbol: Nelfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011914.2 | NP_036044.1 | 10 - 11 | 1429 | 86 | |
| GenBank mRNA | AF101435.1 | - | 10 - 11 | 1381 | 86 | |
| AK019191.1 | - | 10 - 11 | 1495 | 86 | ||
| AK078760.1 | - | 10 - 11 | 1429 | 86 | ||
| AK088384.1 | - | 10 - 11 | 1445 | 86 | ||
| AK144173.1 | - | 1444 | 86 | |||
| AK146185.1 | - | 10 - 11 | 1481 | 86 | ||
| BC018423.1 | - | 1 - 2 | 42 | 86 | ||
| BC038003.1 | - | 10 - 11 | 1430 | 4912719 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of transcription elongation from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
positive regulation of histone H3-K4 methylation
chromatin binding
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of transcription elongation from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
positive regulation of histone H3-K4 methylation
chromatin binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591876 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591874 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591875 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591877 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591878 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591879 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P595954 | BC038003.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P595954 | AK146185.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P595954 | BC018423.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P595954 | AK144173.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P595954 | AK019191.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P595954 | BC038003.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P595954 | AK146185.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P595954 | BC018423.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P595954 | AK144173.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P595954 | AK019191.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 825888 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335825 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303376 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 784264 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406116 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 634147 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300243 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894050 | XM_001474961.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894050 | XM_001476082.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 170534 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 66452 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719752 | AK011451.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719752 | AK138358.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719752 | AY341872.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719752 | AY341878.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719752 | BC052063.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719752 | BC058517.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719752 | NM_028126.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 744500 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148218 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 200205 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336083 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 565622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348893 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 732901 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520501 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 98846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147146 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 68221 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73764 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159770 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48860 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 131964 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 70232 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125315 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113777 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P595954 | BC038003.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P595954 | AK146185.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P595954 | BC018423.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P595954 | AK144173.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P595954 | AK019191.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P595954 | BC038003.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P595954 | AK146185.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P595954 | BC018423.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P595954 | AK144173.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P595954 | AK019191.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P242329 | BC038003.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P242329 | AK146185.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P242329 | AK216954.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P242329 | BC018423.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P242329 | AK144173.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23591885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23591881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23591870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23591871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23591866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23591867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23591883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23591874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23591875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23591868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23591869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23591876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23591864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23591863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23591878 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125315 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05528909 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113777 | - | - |
| Whole Mouse Genome G4122A | A_52_P242329 | BC038003.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P242329 | AK146185.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P242329 | AK216954.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P242329 | BC018423.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P242329 | AK144173.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595954 | BC038003.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595954 | AK146185.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595954 | BC018423.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595954 | AK144173.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595954 | AK019191.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595954 | BC038003.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595954 | AK146185.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595954 | BC018423.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595954 | AK144173.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595954 | AK019191.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105274 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125315 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113777 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04765213 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04973464 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04973453 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04769560 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04973458 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591874 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591878 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591875 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591876 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591879 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23591877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23591858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04563961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04805382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04306467 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04681673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04821949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04348499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23591876 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10529339 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529335 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529338 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529341 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529340 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529342 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529332 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529337 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529334 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529333 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529336 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427248_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10529330 | - | - |
| Mouse Genome 430 2.0 Array | 1427248_at | - | - |
| Mouse Genome 430 2.0 Array | 1427248_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 24116_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 24116_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500020311.mm.1 | NM_011914.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500020311.mm.1 | NM_011914.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500020311.mm.1 | TC0500002223.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427248_at | - | - |
| Mouse Expression Set 430 | 1427248_PM_at | - | - |
| Murine Genome U74v2 Set | 160616_at | - | - |
| Murine Genome U74v2 Set | 160616_PM_at | - | - |
| Murine Genome U74v2 Set | 166174_PM_r_at | - | - |
| Murine Genome U74v2 Set | 134264_PM_at | - | - |
| Murine Genome U74v2 Set | 134264_at | - | - |
| Murine Genome U74v2 Set | 166174_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427248_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427248_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427248_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427248_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106290148 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5900706 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5900706 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106290148 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005900706 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2474887 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2474887 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2465643 | - | - |
| mouse-6 v1.1 (Illumina) | scl0024116.2_231-S | - | - |
| mouse-6 v1.1 (Illumina) | scl26712.11_156-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106290148 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5900706 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0024116.2_231-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106290148 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5900706 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2474887 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2474887 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2474886 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0024116.2_231-S | - | - |

Nelfa
Genomic Map