Assay Details
| Gene Symbol: Ccpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114328.2 | NP_001107800.1 | 6 - 7 | 882 | 82 | |
| NM_001286544.1 | NP_001273473.1 | 6 - 7 | 882 | 82 | ||
| NM_001286545.1 | NP_001273474.1 | 6 - 7 | 882 | 82 | ||
| NM_001286546.1 | NP_001273475.1 | 6 - 7 | 868 | 82 | ||
| NM_028181.5 | NP_082457.3 | 6 - 7 | 882 | 82 | ||
| GenBank mRNA | AK032645.1 | - | 6 - 7 | 754 | 82 | |
| AK076325.2 | - | 6 - 7 | 809 | 82 | ||
| AK128894.1 | - | 6 - 7 | 808 | 82 | ||
| AK134675.1 | - | 6 - 7 | 779 | 82 | ||
| BC043049.1 | - | 6 - 7 | 781 | 5704185 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccpg1
Genomic Map