Assay Details
| Gene Symbol: Tmx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028339.1 | NP_082615.1 | 5 - 6 | 506 | 104 | |
| GenBank mRNA | AK013150.1 | - | 5 - 6 | 506 | 104 | |
| AK030429.1 | - | 5 - 6 | 524 | 104 | ||
| AK050567.1 | - | 5 - 6 | 505 | 104 | ||
| AK137059.1 | - | 5 - 6 | 503 | 104 | ||
| AK150511.1 | - | 5 - 6 | 513 | 104 | ||
| AK153027.1 | - | 5 - 6 | 509 | 104 | ||
| AK170468.1 | - | 5 - 6 | 524 | 104 | ||
| BC017603.1 | - | 5 - 6 | 506 | 4503129 | 104 | |
| BC021533.1 | - | 5 - 6 | 527 | 5354337 | 104 | |
More Information
Set Membership: |
|

Tmx1
Genomic Map