Assay Details
| Gene Symbol: Tmx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028339.1 | NP_082615.1 | 6 - 7 | 606 | 72 | |
| GenBank mRNA | AK013150.1 | - | 6 - 7 | 606 | 72 | |
| AK030429.1 | - | 6 - 7 | 624 | 72 | ||
| AK050567.1 | - | 6 - 7 | 605 | 72 | ||
| AK137059.1 | - | 6 - 7 | 603 | 72 | ||
| AK150511.1 | - | 6 - 7 | 613 | 72 | ||
| AK153027.1 | - | 6 - 7 | 609 | 72 | ||
| AK170468.1 | - | 6 - 7 | 624 | 72 | ||
| BC017603.1 | - | 6 - 7 | 606 | 4503129 | 72 | |
| BC021533.1 | - | 6 - 7 | 627 | 5354337 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmx1
Genomic Map