Assay Details
| Gene Symbol: Klk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174866.2 | NP_777355.1 | 4 - 5 | 602 | 92 | |
| GenBank mRNA | BC044756.2 | - | 3 - 4 | 577 | 92 | |
| BC128019.1 | - | 4 - 5 | 605 | 40136266 | 92 | |
More Information
Set Membership: |
|

Klk14
Genomic Map