Assay Details
| Gene Symbol: Rims3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182929.2 | NP_891559.1 | 3 - 4 | 472 | 78 | |
| GenBank mRNA | AK043152.1 | - | 5 - 6 | 971 | 78 | |
| AK122226.2 | - | 3 - 4 | 340 | 78 | ||
| AY326953.1 | - | 3 - 4 | 472 | 78 | ||
| BC115536.2 | - | 3 - 4 | 472 | 40101461 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rims3
Genomic Map