Assay Details
| Gene Symbol: 4930451I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183131.2 | NP_898954.2 | 2 - 3 | 248 | 71 | |
| GenBank mRNA | AK019625.1 | - | 2 - 3 | 225 | 71 | |
| BC048622.1 | - | 2 - 3 | 248 | 6743640 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

4930451I11Rik
Genomic Map