Assay Details
| Gene Symbol: 4930451I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183131.2 | NP_898954.2 | 3 - 4 | 329 | 67 | |
| GenBank mRNA | AK019625.1 | - | 3 - 4 | 306 | 67 | |
| BC048622.1 | - | 3 - 4 | 329 | 6743640 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412574 | BC048622.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412574 | AK019625.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412574 | NM_183131.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412574 | BC048622.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412574 | AK019625.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412574 | NM_183131.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360875 | AK090348.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360875 | AK136962.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360875 | BC032199.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360875 | NM_025566.2 | refseq_mrna_mouse_NM |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127261 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412574 | BC048622.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412574 | AK019625.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412574 | NM_183131.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412574 | BC048622.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412574 | AK019625.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412574 | NM_183131.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25502519 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127261 | - | - |
| Whole Mouse Genome G4122A | A_52_P412574 | BC048622.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412574 | AK019625.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412574 | NM_183131.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412574 | BC048622.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412574 | AK019625.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412574 | NM_183131.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102130 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110665 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127261 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06305010 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05909602 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05909608 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05909609 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05909605 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06636856 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06467316 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06467360 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06467330 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05909604 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06467355 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06467334 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05909611 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05909612 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06311325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06467338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06311321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06311318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05960659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05909613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05932385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06305010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06636861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05960647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06467335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06467324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06636856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06467330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06221610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06311313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06467327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06311319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06311323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502454 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10568061 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568063 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441086_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10568060 | - | - |
| Mouse Genome 430 2.0 Array | 1441086_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441086_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78118_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78118_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700035991.mm.1 | NM_183131.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035991.mm.1 | NM_183131.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035991.mm.1 | TC0700004300.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1441086_PM_at | - | - |
| Mouse Expression Set 430 | 1441086_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441086_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441086_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 819370 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1940433 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001940433 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2682763 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2682763 | - | - |
| mouse-6 v1.1 (Illumina) | scl30669.5.1_22-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1940433 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30669.5.1_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1940433 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2682763 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2682763 | - | - |

4930451I11Rik
Genomic Map