Assay Details
| Gene Symbol: Fam229b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183254.1 | NP_899077.1 | 1 - 2 | 75 | 80 | |
| GenBank mRNA | AK006353.1 | - | 1 - 2 | 75 | 80 | |
| AK006431.1 | - | 1 - 2 | 56 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006353.1, NM_183254.1 transcripts.
Set Membership: |
|

Fam229b
Genomic Map