Assay Details
| Gene Symbol: Fam229b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183254.1 | NP_899077.1 | 3 - 4 | 360 | 111 | |
| GenBank mRNA | AK006353.1 | - | 3 - 4 | 360 | 111 | |
| AK006431.1 | - | 3 - 4 | 338 | 111 | ||
| BC147171.1 | - | 2 - 3 | 289 | 9056241 | 111 | |
| BC147172.1 | - | 2 - 3 | 289 | 9056208 | 111 | |
| BC172004.1 | - | 2 - 3 | 286 | 9054290 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006353.1, BC147171.1, BC147172.1, BC172004.1, NM_183254.1 transcripts.
Set Membership: |
|

Fam229b
Genomic Map