Assay Details
| Gene Symbol: Fam229b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183254.1 | NP_899077.1 | 4 - 5 | 507 | 76 | |
| GenBank mRNA | AK006353.1 | - | 4 - 5 | 507 | 76 | |
| AK006431.1 | - | 4 - 5 | 486 | 76 | ||
| BC147171.1 | - | 3 - 4 | 436 | 9056241 | 76 | |
| BC147172.1 | - | 3 - 4 | 436 | 9056208 | 76 | |
| BC172004.1 | - | 3 - 4 | 433 | 9054290 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27076705 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27076704 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P651948 | BC094255.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P651948 | BC147171.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P651948 | BC147172.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P651948 | BC172004.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P651948 | NM_183254.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P651948 | BC094255.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P651948 | BC147171.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P651948 | BC147172.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P651948 | BC172004.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P651948 | NM_183254.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 686650 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 920986 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254290 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365405 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 122726 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338513 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 317601 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 308101 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 68700 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11428 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139176 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125226 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P651948 | BC094255.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P651948 | BC147171.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P651948 | BC147172.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P651948 | BC172004.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P651948 | NM_183254.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P651948 | BC094255.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P651948 | BC147171.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P651948 | BC147172.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P651948 | BC172004.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P651948 | NM_183254.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27076695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27076704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27076700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27076690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27076687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27076698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27076703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27076701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27076705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27076697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27076693 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07032985 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139176 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125226 | - | - |
| Whole Mouse Genome G4122A | A_52_P651948 | BC094255.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P651948 | BC147171.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P651948 | BC147172.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P651948 | BC172004.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P651948 | NM_183254.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P651948 | BC094255.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P651948 | BC147171.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P651948 | BC147172.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P651948 | BC172004.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P651948 | NM_183254.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115503 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139176 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125226 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08922099 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08922106 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08922108 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08922101 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27076704 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27076705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27076655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08674662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27076695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08674660 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10368684 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368687 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368685 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368688 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368686 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460553_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10368683 | - | - |
| Mouse Genome 430 2.0 Array | 1460553_at | - | - |
| Mouse Genome 430 2.0 Array | 1460553_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66337_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66337_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000014298.mm.1 | NM_183254.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014298.mm.1 | NM_183254.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014298.mm.1 | TC1000002017.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000014294.mm.1 | NM_183254.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014294.mm.1 | NM_183254.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014294.mm.1 | TC1000002017.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460553_PM_at | - | - |
| Mouse Expression Set 430 | 1460553_at | - | - |
| Murine Genome U74v2 Set | 169895_at | - | - |
| Murine Genome U74v2 Set | 169895_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1460553_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1460553_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 650508 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1230161 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1230161 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001230161 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1214517 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214517 | - | - |
| mouse-6 v1.1 (Illumina) | scl38049.5.1_21-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1230161 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38049.5.1_21-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1230161 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1214517 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1214517 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl38049.5.1_21-S | - | - |

Fam229b
Genomic Map