Assay Details
| Gene Symbol: Fam24a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183272.2 | NP_899095.1 | 2 - 3 | 158 | 84 | |
| GenBank mRNA | AK006815.1 | - | 2 - 3 | 159 | 84 | |
| AK018853.1 | - | 2 - 3 | 160 | 84 | ||
| AK018873.1 | - | 2 - 3 | 198 | 84 | ||
| AK018966.1 | - | 2 - 3 | 198 | 84 | ||
| BC048642.1 | - | 2 - 3 | 215 | 6772199 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P438039 | AK006815.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P438039 | AK018853.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P438039 | AK018873.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P438039 | AK018966.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P438039 | BC048642.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P438039 | AK006815.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P438039 | AK018853.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P438039 | AK018873.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P438039 | AK018966.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P438039 | BC048642.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023364 | AK006815.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023364 | AK018853.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023364 | NM_183272.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 487445 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11446 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134172 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023364 | AK006815.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023364 | AK018853.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023364 | NM_183272.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P438039 | AK006815.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P438039 | AK018853.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P438039 | AK018873.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P438039 | AK018966.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P438039 | BC048642.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P438039 | AK006815.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P438039 | AK018853.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P438039 | AK018873.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P438039 | AK018966.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P438039 | BC048642.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25527873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25527863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25527859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25527860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25527862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25527855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25527856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25527872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25527870 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134172 | - | - |
| Whole Mouse Genome G4122A | A_51_P438039 | AK006815.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438039 | AK018853.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438039 | AK018873.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438039 | AK018966.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438039 | BC048642.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438039 | AK006815.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438039 | AK018853.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438039 | AK018873.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438039 | AK018966.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438039 | BC048642.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112271 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121776 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134172 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06638385 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06048789 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06048788 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06048792 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06048781 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06048793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25527845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05911112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06534920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06501195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06638384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06534919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25527860 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10558217 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558219 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558218 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10558216 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700016832.mm.1 | NM_183272.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016832.mm.1 | NM_183272.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016832.mm.1 | TC0700001918.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016834.mm.1 | NM_183272.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016834.mm.1 | NM_183272.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016834.mm.1 | TC0700001918.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016831.mm.1 | TC0700001918.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016833.mm.1 | NM_183272.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016833.mm.1 | NM_183272.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016833.mm.1 | TC0700001918.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 786851 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 940193 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 940193 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000940193 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1248923 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248923 | - | - |
| mouse-6 v1.1 (Illumina) | scl31979.4.1_56-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 940193 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31979.4.1_56-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 940193 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1248923 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1248923 | - | - |

Fam24a
Genomic Map